N,N-Bis(3-chlorobenzyl)amine ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B1793-25G B1793-5G
TCIAB1793-25GEA 4900 DKK
TCIAB1793-25G TCIAB1793-5G
N,N-Bis(3-chlorobenzyl)amine ≥98.0% (by GC)
N,N-Bis(3-chlorobenzyl)amine
Formel: C₁₄H₁₃Cl₂N
Molekylvægt: 266.17 g/mol
MDL Number: MFCD00671501
CAS-nummer: 129041-31-2

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR