Bis(2-morpholinoethyl)ether ≥85.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4,4'-(Oxybis(ethane-2,1-diyl))dimorpholine, 4,4'-(3-Oxapentane-1,5-diyl)bismorpholine

B1784-25G B1784-500G
TCIAB1784-25GEA 165 DKK
TCIAB1784-25G TCIAB1784-500G
Bis(2-morpholinoethyl)ether ≥85.0% (by GC)
Bis(2-morpholinoethyl)ether
Formel: C₁₂H₂₄N₂O₃
Kogepunkt: 192°C
Flammepunkt: 146°C
MDL Number: MFCD00072740
CAS-nummer: 6425-39-4
EINECS: 229-194-7

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 85.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR