(2-Bromoallyl)trimethylsilane ≥90.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

B1695-1G B1695-5G
TCIAB1695-1GEA 895 DKK
TCIAB1695-1G TCIAB1695-5G
(2-Bromoallyl)trimethylsilane ≥90.0% (by GC)
(2-Bromoallyl)trimethylsilane

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR