Tris(hexafluoroisopropyl)borate ≥95.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Boric Acid Tris(hexafluoroisopropyl) Ester
B1645-1G
B1645-5G
TCIAB1645-1GEA
796
DKK
TCIAB1645-1G
TCIAB1645-5G
Tris(hexafluoroisopropyl)borate ≥95.0% (by GC, titration analysis)
Tris(hexafluoroisopropyl)borate
Formel:
C₉H₃BF₁₈O₃ |
MDL Number:
MFCD00191615 CAS-nummer: 6919-80-8 |
Specifikationer Testresultater
Appearance | White powder to lump |
Purity (GC) | min. 95.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...