Nɑ-(tert-Butoxycarbonyl)-N-ε-carbobenzoxy-L-lysine ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Nepsilon-Benzyloxycarbonyl-Nα-Boc-L-lysine, Boc-Lys(Z)-OH, Nɑ-Boc-Nepsilon-Cbz-L-lysine, Boc-Lys(Cbz)-OH

B1632-25G B1632-5G
TCIAB1632-25GEA 829 DKK
TCIAB1632-25G TCIAB1632-5G
Nɑ-(tert-Butoxycarbonyl)-N-ε-carbobenzoxy-L-lysine ≥98.0% (by HPLC, titration analysis)
Nalpha-(tert-Butoxycarbonyl)-N-epsilon-carbobenzoxy-L-lysine
Formel: C₁₉H₂₈N₂O₆
MDL Number: MFCD00065584
CAS-nummer: 2389-45-9
EINECS: 219-221-0

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 75.0 to 79.0 deg-C
Specific rotation [a]20/D -7.0 to -9.0 deg (C=2.5, AcOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR