(tert-Butyl)ethylether ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
2-Ethoxy-2-methylpropane
B1542-25ML
B1542-5ML
TCIAB1542-25MLEA
427
DKK
TCIAB1542-25ML
TCIAB1542-5ML
(tert-Butyl)ethylether ≥95.0% (by GC)
(tert-Butyl)ethylether
Formel:
C₆H₁₄O Molekylvægt: 102.18 g/mol Kogepunkt: 70°C Smeltepunkt: -97 Flammepunkt: –19°C |
MDL Number:
MFCD00009225 CAS-nummer: 637-92-3 EINECS: 211-309-7 UN nummer: 1179 ADR: 3 II Merck Index: 14,03774 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...