tert-Butyl methyl sulphide ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

B1475-25ML B1475-5ML
TCIAB1475-25MLEA 641 DKK
TCIAB1475-25ML TCIAB1475-5ML
tert-Butyl methyl sulphide ≥98.0% (by GC)
tert-Butyl methyl sulphide
Formel: C₅H₁₂S
Molekylvægt: 104.21 g/mol
Kogepunkt: 102°C
Flammepunkt: –3°C
MDL Number: MFCD00008840
CAS-nummer: 6163-64-0
UN nummer: 1993
ADR: 3 II

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR