2-Bromo-5-nitrofuran ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B1470-1G B1470-5G
TCIAB1470-1GEA 706 DKK
TCIAB1470-1G TCIAB1470-5G
2-Bromo-5-nitrofuran ≥98.0% (by GC)
2-Bromo-5-nitrofuran
Formel: C₄H₂BrNO₃
Molekylvægt: 191.97 g/mol
Kogepunkt: 118°C
MDL Number: MFCD00142879
CAS-nummer: 823-73-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cbc6078a986c1f1d2a928c223ab2fbb4da2a1d41&vid=cbc6078a986c1f1d2a928c223ab2fbb4da2a1d41&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light orange to Yellow to Green powder to crystaline
Purity (GC) min. 98.0 %
Melting point 47.0 to 49.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR