Bis(tetrabutylammonium) bis(1,3-dithiole-2-thione-4,5-dithiolato)palladium(II) ≥90.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

B1437-1G B1437-100MG
TCIAB1437-1GEA 1960 DKK
TCIAB1437-1G TCIAB1437-100MG
Bis(tetrabutylammonium) bis(1,3-dithiole-2-thione-4,5-dithiolato)palladium(II) ≥90.0% (by titrimetric analysis)
Bis(tetrabutylammonium) bis(1,3-dithiole-2-thione-4,5-dithiolato)palladium(II)
Formel: C₃₈H₇₂N₂PdS₁₀
Molekylvægt: 984.03 g/mol
MDL Number: MFCD00143779
CAS-nummer: 72688-90-5

Bestil nu

Specifikationer Testresultater

Appearance Gray to Dark purple to Black powder to crystal
Purity (Chelometric Titration) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR