Bis(dibenzylideneacetone)palladium

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Bis(dibenzylideneacetone)palladium(0)

B1374-1G B1374-5G
TCIAB1374-1GEA 952 DKK
TCIAB1374-1G TCIAB1374-5G
Bis(dibenzylideneacetone)palladium
Bis(dibenzylideneacetone)palladium
Formel: C₃₄H₂₈O₂Pd
MDL Number: MFCD00051942
CAS-nummer: 32005-36-0

Bestil nu

Specifikationer Testresultater

Appearance Red to Dark blue to Black powder to crystal
Content (Palladium) 18.0 to 24.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR