Benzyl Isopropenyl Ether ≥97.0% (by GC) hydroxyl protecting agent

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-Benzyloxy-1-propene, Isopropenyl Benzyl Ether

B1257-1ML B1257-5ML
TCIAB1257-1MLEA 731 DKK
TCIAB1257-1ML TCIAB1257-5ML
Benzyl Isopropenyl Ether ≥97.0% (by GC) hydroxyl protecting agent
Benzyl isopropenyl ether
Formel: C₁₀H₁₂O
Kogepunkt: 192°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00059913
CAS-nummer: 32783-20-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=981d49529e3133dcc2b068e6a0854c609bc50e1b&vid=981d49529e3133dcc2b068e6a0854c609bc50e1b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Yellow to Green clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR