1-Butyl-2-pyrrolidone ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: N-Butyl-2-azacyclopentanone, 1-Butylpyrrolidin-2-one

B1254-25ML B1254-5ML
TCIAB1254-25MLEA 1470 DKK
TCIAB1254-25ML TCIAB1254-5ML
1-Butyl-2-pyrrolidone ≥98.0% (by GC)
1-Butyl-2-pyrrolidone
Formel: C₈H₁₅NO
Molekylvægt: 141.21 g/mol
Kogepunkt: 137°C
MDL Number: MFCD00020876
CAS-nummer: 3470-98-2
EINECS: 222-437-8

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light red to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR