4-Butylbenzaldehyde diethyl acetal ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

B1222-25ML B1222-5ML
TCIAB1222-25MLEA 1730 DKK
TCIAB1222-25ML TCIAB1222-5ML
4-Butylbenzaldehyde diethyl acetal ≥97.0% (by GC)
4-Butylbenzaldehyde diethyl acetal
Formel: C₁₅H₂₄O₂
Molekylvægt: 236.36 g/mol
MDL Number: MFCD00009466
CAS-nummer: 83803-80-9
EINECS: 280-899-6

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR