Bis(tetrabutylammonium) bis(1,3-dithiole-2-thione-4,5-dithiolato)zinc complex ≥98.0% (by total nitrogen and titration analysis) for organic electronics

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: BTBA-BDTD Zn Complex

B1221-25G B1221-5G
TCIAB1221-25GEA 3740 DKK
TCIAB1221-25G TCIAB1221-5G
Bis(tetrabutylammonium) bis(1,3-dithiole-2-thione-4,5-dithiolato)zinc complex ≥98.0% (by total nitrogen and titration analysis) for organic electronics
Bis(tetrabutylammonium) bis(1,3-dithiole-2-thione-4,5-dithiolato)zinc complex
Formel: C₃₈H₇₂N₂S₁₀Zn
Molekylvægt: 942.99 g/mol
MDL Number: MFCD00060267
CAS-nummer: 68449-38-7

Bestil nu

Specifikationer Testresultater

Appearance Red to Dark red to Dark purple powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Chelometric Titration) min. 98.0 %
Melting point 170.0 to 177.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR