4,4'-Bis(dimethylamino)benzil ≥99.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 1,2-Bis(4-(dimethylamino)phenyl)ethane-1,2-dione, 1,2-Bis(4-dimethylaminophenyl)-1,2-ethanedione

B1212-25G B1212-5G
TCIAB1212-25GEA 2720 DKK
TCIAB1212-25G TCIAB1212-5G
4,4'-Bis(dimethylamino)benzil ≥99.0% (by titrimetric analysis)
4,4'-Bis(dimethylamino)benzil
Formel: C₁₈H₂₀N₂O₂
Molekylvægt: 296.37 g/mol
MDL Number: MFCD00051389
CAS-nummer: 17078-27-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af44ca3dd73066343cf559b9549a058c342579b7&vid=af44ca3dd73066343cf559b9549a058c342579b7&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 202.0 to 205.0 deg-C
Lambda max. 370.0 to 372.0 nm (MeOH)
Molar extinction coefficient min. 45,000 (MeOH, 370.0 to 372.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR