Batyl alcohol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

B1202-25G B1202-500G
TCIAB1202-25GEA 411 DKK
TCIAB1202-25G TCIAB1202-500G
Batyl alcohol ≥98.0% (by GC)
Batyl alcohol
Formel: C₂₁H₄₄O₃
MDL Number: MFCD00004720
CAS-nummer: 544-62-7
EINECS: 208-874-7
Merck Index: 14,01011
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a111526501299ed756f7245b15ef1f8408d85ddf&vid=a111526501299ed756f7245b15ef1f8408d85ddf&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 70.0 to 73.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR