4-Bromopyridine hydrobromide ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
B1171-25G
B1171-5G
TCIAB1171-25GEA
2280
DKK
TCIAB1171-25G
TCIAB1171-5G
4-Bromopyridine hydrobromide ≥98.0% (by titrimetric analysis)
4-Bromopyridine hydrobromide
Formel:
C₅H₄BrN Molekylvægt: 238.91 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00060195 CAS-nummer: 74129-11-6 UN nummer: 2811 ADR: 6.1 III |
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...