Benzyl methyl sulphide ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Benzyl(methyl)sulphane

B0992-25ML B0992-5ML
TCIAB0992-25MLEA 944 DKK
TCIAB0992-25ML TCIAB0992-5ML
Benzyl methyl sulphide ≥98.0% (by GC)
Benzyl methyl sulphide
Formel: C₈H₁₀S
Kogepunkt: 200°C
Flammepunkt: 73°C
MDL Number: MFCD00008563
CAS-nummer: 766-92-7
EINECS: 212-174-7

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR