1,1'-Dibenzoylferrocene ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Di(benzoylcyclopentadienyl) Iron, Bis(benzoylcyclopentadienyl) Iron

B0915-25G B0915-5G
TCIAB0915-25GEA 2060 DKK
TCIAB0915-25G TCIAB0915-5G
1,1'-Dibenzoylferrocene ≥98.0% (by HPLC)
1,1'-Dibenzoylferrocene
Formel: C₂₄H₁₀FeO₂
Molekylvægt: 386.18 g/mol
MDL Number: MFCD00045455
CAS-nummer: 12180-80-2
EINECS: 235-344-2

Bestil nu

Specifikationer Testresultater

Appearance Orange to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 102.0 to 106.0 deg-C
Solubility in Toluene almost transparency

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR