N-tert-Butylethylamine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

B0888-25ML B0888-5ML
TCIAB0888-25MLEA 1220 DKK
TCIAB0888-25ML TCIAB0888-5ML
N-tert-Butylethylamine ≥98.0% (by GC, titration analysis)
N-tert-Butylethylamine
Formel: C₆H₁₅N
Kogepunkt: 89°C
MDL Number: MFCD00059367
CAS-nummer: 4432-77-3
UN nummer: 2733
ADR: 3,II

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR