O-Benzyl-DL-serine ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
B0860-1G
B0860-10G
TCIAB0860-1GEA
148
DKK
TCIAB0860-1G
TCIAB0860-10G
O-Benzyl-DL-serine ≥98.0% (by titrimetric analysis)
O-Benzyl-DL-serine
Formel:
C₁₀H₁₃NO₃ |
MDL Number:
MFCD00021724 CAS-nummer: 5445-44-3 EINECS: 226-650-7 |
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in hot Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...