Bis(chloromethyl)sulphide ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

B0843-25G B0843-5G B0843-1G
TCIAB0843-25GEA 3980 DKK
TCIAB0843-25G TCIAB0843-5G TCIAB0843-1G
Bis(chloromethyl)sulphide ≥97.0% (by GC)
Bis(chloromethyl)sulphide
Formel: C₂H₄Cl₂S
Kogepunkt: 156°C
MDL Number: MFCD00059474
CAS-nummer: 3592-44-7
UN nummer: 1992
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b984b105076c6c3fbe9871997a3e6bbfc8bf48e9&vid=b984b105076c6c3fbe9871997a3e6bbfc8bf48e9&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR