Amyl butyrate 98,0 GC_ASSAY_METHOD

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Butyric Acid Amyl Ester, Pentyl Butyrate

B0755-25ML B0755-500ML
TCIAB0755-25MLEA 132 DKK
TCIAB0755-25ML TCIAB0755-500ML
Amyl butyrate 98,0 GC_ASSAY_METHOD
Amyl butyrate
Formel: C₉H₁₈O₂
Molekylvægt: 158.24 g/mol
Flammepunkt: 57°C
MDL Number: MFCD00048850
CAS-nummer: 540-18-1
EINECS: 208-739-2
UN nummer: 2620
ADR: 3 III
Merck Index: 14,00604

Bestil nu

Specifikationer Testresultater

Appearance Colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR