Tetrabutyl orthotitanate tetramer
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Butyl Titanate Tetramer, Tetrabutoxy Titan Tetramer, Tetrabutyl Titanate Tetramer, Titanium Butoxide Tetramer
B0743-25G
B0743-100G
TCIAB0743-25GEA
189
DKK
TCIAB0743-25G
TCIAB0743-100G
Tetrabutyl orthotitanate tetramer
Tetrabutyl orthotitanate tetramer
Formel:
C₁₆H₃₆O₄Ti Flammepunkt: 50°C |
MDL Number:
MFCD00070567 CAS-nummer: 70799-68-7 UN nummer: 1993 ADR: 3,III |
Specifikationer Testresultater
Appearance | Colorless to Yellow clear liquid |
Ignition residue | 31.3 to 33.6 % (as TiO2) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...