4,4'-Butylidenebis(6-tert-butyl-m-cresol) ≥97.0% (by GC)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: 4,4'-(Butane-1,1-diyl)bis(2-(tert-butyl)-5-methylphenol)
B0724-25G B0724-500G
TCIAB0724-25GEA 132 DKK
TCIAB0724-25G TCIAB0724-500G
4,4'-Butylidenebis(6-tert-butyl-m-cresol) ≥97.0% (by GC)
4,4'-Butylidenebis(6-tert-butyl-m-cresol)
Formel: C₂₆H₃₈O₂
Molekylvægt: 382.59 g/mol
MDL Number: MFCD00026289
CAS-nummer: 85-60-9
EINECS: 201-618-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9814edef4390ebb8b4d26ec5070253de68395237&vid=9814edef4390ebb8b4d26ec5070253de68395237&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 208.0 to 213.0 deg-C
Solubility in Methanol almost transparency