Beryllon III

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 5-(4-Diethylamino-2-hydroxyphenylazo)-4-hydroxynaphthalene-2,7-disulphonic Acid Sodium Salt

B0453-1G B0453-5G
TCIAB0453-1GEA 378 DKK
TCIAB0453-1G TCIAB0453-5G
Beryllon III
Beryllon III
Formel: C₂₀H₂₁N₃O₈S₂
Molekylvægt: 517.50 g/mol
MDL Number: MFCD00070629
CAS-nummer: 3627-04-1

Bestil nu

Specifikationer Testresultater

Appearance Dark green to Dark blue to Black powder to crystal
suitability for Berylium-analysis molar extinction coefficient >15000 (pH12.4 , 523.0 to 529.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR