Benzyl orange

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

B0425-1G B0425-5G
TCIAB0425-1GEA 501 DKK
TCIAB0425-1G TCIAB0425-5G
Benzyl orange
Benzyl orange
Formel: C₁₉H₁₇N₃O₃S
MDL Number: MFCD00198097
CAS-nummer: 36402-77-4

Bestil nu

Specifikationer Testresultater

Appearance Orange to Brown powder to crystal
Lambda max. 432.0 to 438.0 nm (H2O)
Absorbance (E1%1cm) min. 550 (H2O, 432.0 to 438.0 nm)
Transition interval (pH) (red)1.9 to 3.3 (redish yellow)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR