2-Ethylhexyl butyrate ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Butyric acid 2-ethylhexyl ester, Butyric acid octyl ester
B0130-25ML
B0130-500ML
TCIAB0130-25MLEA
263
DKK
TCIAB0130-25ML
TCIAB0130-500ML
2-Ethylhexyl butyrate ≥98.0% (by GC)
2-Ethylhexyl butyrate
Formel:
C₁₂H₂₄O₂ Molekylvægt: 200.32 g/mol Kogepunkt: 237°C |
MDL Number:
MFCD00059428 CAS-nummer: 25415-84-3 EINECS: 246-959-0 |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...