Benzo[h]quinoline ≥99.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

B0088-25G B0088-5G
TCIAB0088-25GEA 1570 DKK
TCIAB0088-25G TCIAB0088-5G
Benzo[h]quinoline ≥99.0% (by GC, titration analysis)
Benzo[h]quinoline
Formel: C₁₃H₉N
Kogepunkt: 338°C
MDL Number: MFCD00004984
CAS-nummer: 230-27-3
EINECS: 205-937-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22f40142cb0ec1f07e08893d4646db82ba24abb6&vid=22f40142cb0ec1f07e08893d4646db82ba24abb6&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 50.0 to 54.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR