Undecafluorohexanoic acid ≥98.0% (by titrimetric analysis), High Grade til ion-par kromatografi, til LC-MS
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
A5722-1G
A5722-5G
TCIAA5722-1GEA
559
DKK
TCIAA5722-1G
TCIAA5722-5G
Undecafluorohexanoic acid ≥98.0% (by titrimetric analysis), High Grade til ion-par kromatografi, til LC-MS
Undecafluorohexanoic acid
Formel:
C₆HF₁₁O₂ Kogepunkt: 157°C Smeltepunkt: 14°C Massefylde: 1,76 |
MDL Number:
MFCD00198040 CAS-nummer: 307-24-4 EINECS: 206-196-6 UN nummer: 3265 ADR: 8,II |
Specifikationer Testresultater
Appearance | White or Colorless to Almost white or Almost colorless powder to lump to clear liquid |
Purity (Neutralization titration) | min. 98.0 % |
Absorbance (230nm) | max. 0.20 |
Absorbance (254nm) | max. 0.01 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...