Dihexylammonium acetate ∼0.5 mol/L in water til ion-par kromatografi, til LC-MS

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

A5705-10ML A5705-100ML
TCIAA5705-10MLEA 181 DKK
TCIAA5705-10ML TCIAA5705-100ML
Dihexylammonium acetate ∼0.5 mol/L in water til ion-par kromatografi, til LC-MS
Dihexylammonium acetate
Formel: C₁₄H₃₁NO₂
MDL Number: MFCD03093619
CAS-nummer: 366793-17-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab3add1c37c75b0e2ff9212dbe59628109db2afa&vid=a9c7ed42a241f97d0421c51d4aacc91d1c305fd7&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless clear liquid
Concentration (Nonaqueous Titration) 0.50 to 0.55 mol/L
Absorbance (230nm) max. 2.00
Absorbance (254nm) max. 0.20
Absorbance (270nm) max. 0.20


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR