Malonamid ≥98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A17939.0E
A17939.22
A17939.36
A17939.0EEA
8140
DKK
A17939.0E
A17939.22
A17939.36
Malonamid ≥98%
Malonamid
Purity: 98%
CAS No.: 108-13-4
Molecular Formula: C3H6N2O2
Formula Weight: 102.09
Melting Point: 167-171°
Beilstein Reference: 1751401
Formel:
C₃H₆N₂O₂ Kogepunkt: 461 °C (1013 hPa) Smeltepunkt: 167…171 °C Massefylde: 1,43 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00008034 CAS-nummer: 108-13-4 EINECS: 203-553-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...