Diamylammonium acetate ∼0.5 mol/L in water til ion-par kromatografi, til LC-MS
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A5704-100ML
A5704-10ML
TCIAA5704-100MLEA
1450
DKK
TCIAA5704-100ML
TCIAA5704-10ML
Diamylammonium acetate ∼0.5 mol/L in water til ion-par kromatografi, til LC-MS
Diamylammonium acetate
Formel:
C₁₂H₂₇NO₂ |
MDL Number:
MFCD03093618 CAS-nummer: 211676-91-4 |
|
Specifikationer Testresultater
Appearance | Colorless clear liquid |
Concentration (Nonaqueous Titration) | 0.50 to 0.55 mol/L |
Absorbance (230nm) | max. 2.000 |
Absorbance (254nm) | max. 0.200 |
Absorbance (270nm) | max. 0.200 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...