2,3,4,6-Tetra-O-acetyl-β-D-glucopyranosyl isothiocyanate 98,0 HPLC, N HPLC Labeling

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: (2R,3R,4S,5R,6R)-2-(Acetoxymethyl)-6-isothiocyanatotetrahydro-2H-pyran-3,4,5-triyl triacetate, GITC

A5514-1G A5514-100MG
TCIAA5514-1GEA 2230 DKK
TCIAA5514-1G TCIAA5514-100MG
2,3,4,6-Tetra-O-acetyl-β-D-glucopyranosyl isothiocyanate 98,0 HPLC, N HPLC Labeling
2,3,4,6-Tetra-O-acetyl-beta-D-glucopyranosyl isothiocyanate
Formel: C₁₅H₁₉NO₉S
Molekylvægt: 389.38 g/mol
Smeltepunkt: 116°C
MDL Number: MFCD00043085
CAS-nummer: 14152-97-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9dc5511bc38e425b0e6740367a4cbed01654c80b&vid=9dc5511bc38e425b0e6740367a4cbed01654c80b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 113.0 to 117.0 deg-C
Specific rotation [a]20/D -2.0 to -5.0 deg (C=1,methylene chloride)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR