Sulphonazo III sodium salt ≥90.0% (by HPLC) for spectrophotometry

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2,7-Bis(2-sulphophenylazo)-1,8-dihydroxynaphthalene-3,6-disulphonic Acid Tetrasodium Salt

A5010-1G A5010-5G
TCIAA5010-1GEA 591 DKK
TCIAA5010-1G TCIAA5010-5G
Sulphonazo III sodium salt ≥90.0% (by HPLC) for spectrophotometry
Sulphonazo III sodium salt

Spectrophotometric reagent for alkaline earth metals and indicator for the precipitation titration of SO4 with Ba

Formel: C₂₂H₁₆N₄O₁₄S₄
Molekylvægt: 776.55 g/mol
MDL Number: MFCD00003942
CAS-nummer: 68504-35-8
EINECS: 270-924-9

Bestil nu

Specifikationer Testresultater

Appearance Dark green to Black powder to crystal
Purity (HPLC) min. 90.0 area%
Sensitiveness Colorimetric Test : Turn Purple with Barium


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR