Astragaloside IV ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (2R,3R,4S,5S,6R)-2-(((2aR,3R,4S,5aS,5bS,7S,7aR,9S,11aR,12aS)-4-Hydroxy-3-((2R,5S)-5-(2-hydroxypropan-2-yl)-2-methyltetrahydrofuran-2-yl)-2a,5a,8,8-tetramethyl-9-(((2S,3R,4S,5R)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)oxy)tetradecahydro-1H,12H-cyclopenta[a]cyclopropa[e]phenanthren-7-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

A3305-1G A3305-250MG
TCIAA3305-1GEA 2710 DKK
TCIAA3305-1G TCIAA3305-250MG
Astragaloside IV ≥98.0% (by HPLC)
Astragaloside IV
Formel: C₄₁H₆₈O₁₄
Opbevaringstemperatur: Fryser
MDL Number: MFCD16036240
CAS-nummer: 84687-43-4

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) (CAD) min. 98.0 area%
Specific rotation [a]20/D +19.0 to +26.0 deg (C=0.2, MeOH)
Water max. 7.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR