Azetidin ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

A3257-1G A3257-5G
TCIAA3257-1GEA 641 DKK
TCIAA3257-1G TCIAA3257-5G
Azetidin ≥98.0% (by GC)
Azetidin
Formel: C₃H₇N
Kogepunkt: 62°C
Flammepunkt: -21°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00005165
CAS-nummer: 503-29-7
EINECS: 207-963-8
UN nummer: 2733
ADR: 3,II

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR