Acrylic anhydride ≥95.0% (by GC, titration analysis) stabilised

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A3243-1G A3243-5G
TCIAA3243-1GEA 230 DKK
TCIAA3243-1G TCIAA3243-5G
Acrylic anhydride ≥95.0% (by GC, titration analysis) stabilised
Acrylic anhydride
Formel: C₆H₆O₃
Molekylvægt: 126.11 g/mol
Kogepunkt: 67°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00048146
CAS-nummer: 2051-76-5
EINECS: 218-128-2
UN nummer: 3265
ADR: 8 II

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %
Purity (Morpholine Method) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR