AGK2 ≥97.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A3193-5MG A3193-25MG A3193-100MG
TCIAA3193-5MGEA 517 DKK
TCIAA3193-5MG TCIAA3193-25MG TCIAA3193-100MG
AGK2 ≥97.0% (by HPLC)
AGK2
Formel: C₂₃H₁₃Cl₂N₃O₂
Opbevaringstemperatur: Køleskab
MDL Number: MFCD01909444
CAS-nummer: 304896-28-4

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 97.0 area%
Elemental analysis (Nitrogen) 9.40 to 10.20 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR