10-Acetylphenothiazine ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 1-(10H-Phenothiazin-10-yl)ethanone, 10-Acetyl-10H-phenothiazine

A3154-1G A3154-5G
TCIAA3154-1GEA 698 DKK
TCIAA3154-1G TCIAA3154-5G
10-Acetylphenothiazine ≥98.0% (by GC)
10-Acetylphenothiazine
Formel: C₁₄H₁₁NOS
Molekylvægt: 241.31 g/mol
MDL Number: MFCD00022264
CAS-nummer: 1628-29-1
EINECS: 216-621-7

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 202.0 to 206.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR