5-Azoniaspiro[4.4]nonane tetrafluoroborate ≥98.0% (by total nitrogen basis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

A3103-1G A3103-5G
TCIAA3103-1GEA 378 DKK
TCIAA3103-1G TCIAA3103-5G
5-Azoniaspiro[4.4]nonane tetrafluoroborate ≥98.0% (by total nitrogen basis)
5-Azoniaspiro[4.4]nonane tetrafluoroborate
Formel: C₈H₁₆BF₄N
Molekylvægt: 213.03 g/mol
CAS-nummer: 129211-47-8
UN nummer: 1759
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=330f1673f9e5ce26bfa7771fac75219cd25b0a04&vid=c9a6b1e841840bda4e45bc3715accac9bdecd456&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 188.0 to 192.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR