5-Azoniaspiro[4.4]nonane bis(trifluoromethanesulphonyl)imide ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A3101-1G A3101-5G
TCIAA3101-1GEA 550 DKK
TCIAA3101-1G TCIAA3101-5G
5-Azoniaspiro[4.4]nonane bis(trifluoromethanesulphonyl)imide ≥98.0% (by titrimetric analysis)
5-Azoniaspiro[4.4]nonane bis(trifluoromethanesulphonyl)imide
Formel: C₁₀H₁₆F₆N₂O₄S₂
Molekylvægt: 406.36 g/mol
CAS-nummer: 706759-44-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e7800d6b2fa7fc64f422daae40dcc45a62e10b1f&vid=e7800d6b2fa7fc64f422daae40dcc45a62e10b1f&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 98.0 %
Melting point 112.0 to 116.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR