Ammoniumthiocyanat ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Thiocyanic acid ammonium salt, Ammonium rhodanide, Ammonium sulphocyanate

A3088-25G A3088-500G
TCIAA3088-25GEA 132 DKK
TCIAA3088-25G TCIAA3088-500G
Ammoniumthiocyanat ≥98.0% (by titrimetric analysis)
Ammonium thiocyanate
Formel: NH₄SCN
Molekylvægt: 76.12 g/mol
Smeltepunkt: 150°C
MDL Number: MFCD00011428
CAS-nummer: 1762-95-4
EINECS: 217-175-6
Merck Index: 14,00561
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65f3281f4ccf70636fad690fddca809139665161&vid=65f3281f4ccf70636fad690fddca809139665161&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (NH4SCN method) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR