Aniline hydrobromide ≥98.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Anilinium bromide, Aniline Hydrobromide, Anilinium bromide, Benzenamine hydrobromide, Benzenaminium bromide, Greatcell Solar
A2985-1G
A2985-5G
TCIAA2985-1GEA
296
DKK
TCIAA2985-1G
TCIAA2985-5G
Aniline hydrobromide ≥98.0% (by HPLC)
Aniline hydrobromide
Formel:
C₆H₇N·HBr Smeltepunkt: 285°C |
MDL Number:
MFCD00035465 CAS-nummer: 542-11-0 EINECS: 208-801-9 |
|
Specifikationer Testresultater
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Argentmetric Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...