(3R)-3-Aminobutan-1-ol ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
1-Butanol,3-aMino-, (3R)-, (R)-3-Aminobutanol, (R)-3-Amino-1-butanol, (R)-3-Aminobutan-1-ol, (-)-(R)-3-aMinopropan-1-ol, (3R)-3-Amino-1-butanol, R-3-amino-1-butanol
A2951-1G
A2951-5G
TCIAA2951-1GEA
394
DKK
TCIAA2951-1G
TCIAA2951-5G
(3R)-3-Aminobutan-1-ol ≥98.0% (by GC, titration analysis)
(3R)-3-Aminobutan-1-ol
Formel:
C₄H₁₁NO |
MDL Number:
MFCD13184351 CAS-nummer: 61477-40-5 UN nummer: 2734 ADR: 8,II |
Specifikationer Testresultater
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | -12.0 to -10.0 deg (C=1,EtOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...