1-Acetylisatin ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-Acetylindoline-2,3-dione

A2941-5G A2941-25G
TCIAA2941-5GEA 649 DKK
TCIAA2941-5G TCIAA2941-25G
1-Acetylisatin ≥98.0% (by GC)
1-Acetylisatin
Formel: C₁₀H₇NO₃
Molekylvægt: 189.17 g/mol
MDL Number: MFCD00158542
CAS-nummer: 574-17-4
EINECS: 209-368-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77292d946d9397c8c8acc498eddcdcd81ead2278&vid=dd19f4dccb0cf51405782731edcd9732bc957128&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 141.0 to 145.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR