Fmoc-Glu-OAll ≥96.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: N-Fmoc-L-glutamic acid 1-allyl ester, Fmoc-Glu-allyl ester, Fmoc-Glu-OAll, 1-Allyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamate, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamic acid 1-Allyl ester, 1-Allyl N-Fmoc-L-glutamate, N-Fmoc-L-glutamic acid 1-Allyl ester

A2909-5G A2909-1G
TCIAA2909-5GEA 1420 DKK
TCIAA2909-5G TCIAA2909-1G
Fmoc-Glu-OAll ≥96.0% (by HPLC, titration analysis)
Fmoc-Glu-OAll
Formel: C₂₃H₂₃NO₆
Molekylvægt: 409.438 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00467718
CAS-nummer: 144120-54-7

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Neutralization titration) min. 96.0 %
Melting point 117.0 to 121.0 deg-C
Specific rotation [a]20/D -17.0 to -21.0 deg (C=1,DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR