2-Amino-4-(1-naphthyl)thiazole ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A2888-1G A2888-5G
TCIAA2888-1GEA 329 DKK
TCIAA2888-1G TCIAA2888-5G
2-Amino-4-(1-naphthyl)thiazole ≥97.0% (by GC, titration analysis)
2-Amino-4-(1-naphthyl)thiazole
Formel: C₁₃H₁₀N₂S
MDL Number: MFCD00236016
CAS-nummer: 56503-96-9

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 156.0 to 160.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR