5-Acetyl-2-amino-4-methylthiazole ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-(2-Amino-4-methylthiazol-5-yl)ethanone

A2887-1G A2887-5G
TCIAA2887-1GEA 189 DKK
TCIAA2887-1G TCIAA2887-5G
5-Acetyl-2-amino-4-methylthiazole ≥98.0% (by GC, titration analysis)
5-Acetyl-2-amino-4-methylthiazole
Formel: C₆H₈N₂OS
Molekylvægt: 156.21 g/mol
Smeltepunkt: 266°C
MDL Number: MFCD00051952
CAS-nummer: 30748-47-1

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR