4-Hydroxy-m-anisidine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Amino-2-methoxyphenol, 4-Aminoguaiacol, 4-Hydroxy-3-methoxyaniline

A2883-1G A2883-5G
TCIAA2883-1GEA 517 DKK
TCIAA2883-1G TCIAA2883-5G
4-Hydroxy-m-anisidine ≥98.0% (by GC, titration analysis)
4-Hydroxy-m-anisidine
Formel: C₇H₉NO₂
MDL Number: MFCD01707809
CAS-nummer: 52200-90-5

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 178.0 to 182.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR